ContactUsLinkComponent
NEB®10-beta Electrocompetent E.coli, New England Biolabs
NEB®10-beta Electrocompetent E.coli, New England Biolabs
Catalog # 101417-922
Small Business Enterprise
undefined
NEB®10-beta Electrocompetent E.coli, New England Biolabs
Catalog # 101417-922
Supplier Number:  C3020K
Small Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    NEB 10-beta electrocompetent E.coli
  • Size:
    6×0.1 ml
  • Cat. No.:
    101417-922
  • Supplier no.:
    C3020K
Specifications
About this item
NEB 10-beta electrocompetent E. coli cells are optimized for high efficiency transformation by electroporation. These cells are ideal for DNA library constructions and all cloning purposes.
  • High efficiency strain ideal for cloning large plasmids and BACs
  • Also available in chemically-competent format (single-use and 200 µl vials)
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
  • Free of animal products