ContactUsLinkComponent
microRNA Marker, New England Biolabs
microRNA Marker, New England Biolabs
Catalog # 101418-106
Small Business Enterprise
CAS Number:  
undefined
microRNA Marker, New England Biolabs
Catalog # 101418-106
Supplier Number:  N2102S
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    microRNA Marker
  • Size:
    100 gel lanes
  • Cat. No.:
    101418-106
  • Supplier no.:
    N2102S
Specifications
About this item
The microRNA Marker is a set of three synthetic single-stranded RNA oligonucleotides 17, 21 and 25 residues long that have free 5´ ends (i.e., no 5´ phosphate groups)
These oligonucleotides can be used as size markers on denaturing polyacrylamide gels and Northern blots. The microRNA Marker is best visualized by staining with SYBR-Gold instead of ethidium bromide. The three marker oligos contain the same core sequence so they can be detected by hybridization with the same probe.