ContactUsLinkComponent
Lambda DNA-Mono Cut Mix, New England Biolabs
Lambda DNA-Mono Cut Mix, New England Biolabs
Catalog # 101228-448
Small Business Enterprise
CAS Number:  
undefined
Lambda DNA-Mono Cut Mix, New England Biolabs
Catalog # 101228-448
Supplier Number:  N3019S
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    Lambda DNA-Mono Cut Mix
  • Size:
    100 gel lanes
  • Cat. No.:
    101228-448
  • Supplier no.:
    N3019S
Specifications
About this item
The Mono Cut Mix is a mixture of intact lambda DNA and lambda DNA separately digested with ApaI, KpnI, XbaI and XhoI
The fragments have been filled in with DNA Polymerase I Large (Klenow) Fragment to prevent re-annealing. These fragments are best separated by Pulsed Field Gel Electrophoresis.The approximate mass of DNA in each of the bands is provided (assuming a 0.5 μg load) for approximating the mass of DNA in comparably intense samples of similar size. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.