Specifications
- Size:1000 units
- Enzyme Name:Klenow Fragment (3'-5' exo-)
- Enzyme Activity:50000 u/ml
- Cat. No.:101228-220
- Supplier no.:M0212M
Specifications
About this item
Klenow Fragment (3´→ 5´ exo-) is an N-terminal truncation of DNA Polymerase I which retains polymerase activity, but has lost the 5´→ 3´ exonuclease activity and has mutations (D355A, E357A) which abolish the 3´→ 5´ exonuclease activity.