To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Application:
- Description:Gibson Assembly Cloning Kit
- Environmentally Preferable:
- Cat. No.:102715-912
- No. of reactions:10
- Supplier no.:E5510S
The Gibson Assembly Cloning Kit combines the power of the Gibson Assembly Master Mix with NEB 5-alpha Competent E.coli, enabling successful assembly of multiple DNA fragments, regardless of fragment length or end compatibility and transformation in just under 2 hours.
The Gibson Assembly Cloning Kit has been optimized for the assembly and cloning of up to 6 fragments. It has been rapidly adopted by the synthetic biology community due to its ease-of-use, flexibility and suitability for large DNA constructs.