ContactUsLinkComponent
Gel Loading Dye Blue (6X), New England Biolabs
Gel Loading Dye Blue (6X), New England Biolabs
Catalog # 101417-880
Small Business Enterprise
CAS Number:  
undefined
Gel Loading Dye Blue (6X), New England Biolabs
Catalog # 101417-880
Supplier Number:  B7021S
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    Gel Loading Dye Blue (6X)
  • Size:
    6.0 mL
  • Cat. No.:
    101417-880
  • Supplier no.:
    B7021S
Specifications
About this item
Gel Loading Dye, Blue (6X) is a pre-mixed loading buffer with one tracking dye for agarose and non-denaturing polyacrylamide gel electrophoresis
This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to DNA following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. Bromophenol blue is the standard tracking dye for electrophoresis. It migrates at approximately 300 bp on a standard 1% TBE agarose gel.