Order Entry
United States
ContactUsLinkComponent
EPIMARK® Bisulfite Conversion Kit, New England Biolabs
EPIMARK® Bisulfite Conversion Kit, New England Biolabs
Catalog # 101710-276
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
undefined
EPIMARK® Bisulfite Conversion Kit, New England Biolabs
Catalog # 101710-276
Supplier:  Avantor
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Methylated DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    EpiMark Bisulfite Conversion Kit
  • Size:
    48 Reactions
  • Environmentally Preferable:
  • Description:
    EpiMark Bisulfite Conversion Kit
  • Cat. No.:
    101710-276
  • Supplier no.:
    E3318S

Specifications

About this item

This technique can reveal the methylation status of every cytosine residue, and it is amenable to massively parallel sequencing methods.

Bisulfite conversion involves the conversion of unmodified cytosines to uracil, leaving the modified bases 5-mC and 5-hmC. The EpiMark Bisulfite Conversion Kit is designed for the detection of methylated cytosine, using a series of alternating cycles of thermal denaturation, followed by incubation with sodium bisulfite. This kit includes all the reagents necessary for complete bisulfite conversion, including spin columns. Amplification of bisulfite-treated samples can then be performed using EpiMark Hot Start Taq DNA Polymerase. This kit provides materials for 48 reactions. For optimal results, 50 ng–2 μg DNA is recommended.