CpG Methyltransferase (M.SssI)
The CpG Methyltransferase (M.SssI) methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5'...CG...3'.
The CpG Methyltransferase (M.SssI) methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5'...CG...3'.
- Catalog No:
- 101228-264
- 101228-260
- Enzyme Name:
- CpG Methyltransferase (M.SssI)
- CpG Methyltransferase (M.SssI)
- Concentration:
- 4000 u/ml
- 4000 u/ml
Product Family Options
- Size
- : M0226SSmall Business Enterprise
Specifications
Cat. No.101228-264Enzyme NameCpG Methyltransferase (M.SssI)Concentration4000 u/mlSmall Business Enterprise - : M0226LSmall Business Enterprise
Specifications
Cat. No.101228-260Enzyme NameCpG Methyltransferase (M.SssI)Concentration4000 u/mlSmall Business Enterprise