Casein Kinase II
About this item
Casein Kinase II (CK2) is a constitutively active serine/threonine protein kinase composed of two 44 kDa catalytic α-subunits and two 26 kDa regulatory β-subunits in an α2β2 configuration to form stable heterotetramers
CK2 holoenzyme undergoes autophosphorylation at two serine residues (S2/S3) of its β-subunit. Recently it has been shown that CK2 α-subunits undergo intermolecular tyrosine-autophosphorylation at Y182, which may represent a specific regulatory mechanism. Also, CK2 is able to phosphorylate, under special circumstances, tyrosyl residues in proteins. CK2 is implicated in a variety of cellular functions.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Casein Kinase II (CK2) is a constitutively active serine/threonine protein kinase composed of two 44 kDa catalytic α-subunits and two 26 kDa regulatory β-subunits in an α2β2 configuration to form stable heterotetramers
CK2 holoenzyme undergoes autophosphorylation at two serine residues (S2/S3) of its β-subunit. Recently it has been shown that CK2 α-subunits undergo intermolecular tyrosine-autophosphorylation at Y182, which may represent a specific regulatory mechanism. Also, CK2 is able to phosphorylate, under special circumstances, tyrosyl residues in proteins. CK2 is implicated in a variety of cellular functions.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 101641-528
- 101641-526
- Enzyme Name:
- Casein Kinase II
- Casein Kinase II
- Enzyme Synonyms:
- CKII
- CKII
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: P6010SItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Small Business Enterprise
More
Specifications
Cat. No.101641-528Enzyme NameCasein Kinase IIEnzyme SynonymsCKIISmall Business Enterprise - Supplier Number: P6010LItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Small Business Enterprise
More
Specifications
Cat. No.101641-526Enzyme NameCasein Kinase IIEnzyme SynonymsCKIISmall Business Enterprise