Blue Protein Loading Dye, New England Biolabs
Catalog # 101227-470
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Specifications
- Description:Blue protein loading dye
- Size:8.0 ml
- Cat. No.:101227-470
- Supplier no.:B7703S
Specifications
About this item
The Blue Loading Buffer Pack contains 3X Blue Loading Buffer (8ml) and 30X Reducing Agent (1ml).