Orders LinkContactUsLinkComponent
beta-1-3 Galactosidase
beta-1-3 Galactosidase
500 units
Catalog # 101227-738
Small Business Enterprise
Supplier #:  P0726S
beta-1-3 Galactosidase
Catalog # 101227-738
Small Business Enterprise
CAS Number:  
beta-1-3 Galactosidase
beta-1-3 Galactosidase
500 units
CAS Number:  
Price
$165.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
β1-3 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of β1-3 and, at a much lower rate, β1-6 linked D-galactopyranosyl residues from oligosaccharides
The approximate kinetic data show > 100-fold preference for β1-3 over β1-6 linkages, and > 500-fold preference from β1-3 over β1-4 linkages.
Specifications
Specifications
  • Size:
    500 units
  • Cat. No.:
    101227-738
  • Enzyme Name:
    beta-1-3 Galactosidase