ContactUsLinkComponent
96-Well Microtiter Plate Magnetic Separation Rack, New England Biolabs
96-Well Microtiter Plate Magnetic Separation Rack, New England Biolabs
Catalog # 101641-216
Small Business Enterprise
undefined
96-Well Microtiter Plate Magnetic Separation Rack, New England Biolabs
Catalog # 101641-216
Supplier Number:  S1511S
Small Business Enterprise
Specifications
  • Description:
    96-well Microtiter Plate Magnetic Separation Rack
  • Cat. No.:
    101641-216
  • Supplier no.:
    S1511S
Specifications
About this item
The 96-well plate magnetic rack is designed to be used with commercially available high flanged 100 µl to 300 µl flat-bottom 96-well microplates
24 side-pull magnetic pins attract magnetic beads from solution to the side walls of four adjacent wells. The orientation of the magnetic field ensures complete removal of the magnetic beads from solution during pipetting steps, minimizing sample loss.