Specifications
- Description:9 Degrees North DNA Ligase
- Size:2,500 units
- Cat. No.:200064-420
- Supplier no.:M0238S
Specifications
About this item
9°N™ DNA Ligase catalyzes the formation of a phosphodiester bond between juxtaposed 5´ phosphate and 3´ hydroxyl termini of two adjacent oligonucleotides which are hybridized to a complementary target DNA
9°N DNA Ligase is active at elevated temperatures (45°C-90°C).