Anti-Climb Adapter, Glassco
Catalog # 89426-392
Supplier: Glassco
Specifications
- Capacity:250 ml
- Description:Anti-Climb Adapter
- Cat. No.:89426-392
- Bottom [ST] joint size:24/40
- Top [ST] joint size:[ST] 24/40
- Supplier no.:030.476.02
Specifications
About this item
The trap of this adapter provides expanded volume for reactions that have a tendency to foam excessively.
- Sealed inner vapor tube
- Manufactured from borosilicate 3.3 glass
The inner vapor tube is sealed to the top of the trap to prevent entry into condenser.