Orders LinkContactUsLinkComponent
Erlenmeyer Flask, Glassco
 :   Glassco
Erlenmeyer Flask, Glassco
 :  Glassco
Erlenmeyer Flask, Glassco
Erlenmeyer Flask, Glassco
Manufactured from Borosilicate 3.3 glass.
Stopper is not supplied.
Product Family Options
  • Capacity
    [ST] joint no.
  • 10 ml
    14/20
    Each
    Each - $20.75
    $20.75
    Each
       $20.75
     : 071.470.04
  • 25 ml
    14/20
    Each
    Each - $20.75
    $20.75
    Each
       $20.75
     : 071.470.06
  • 50 ml
    14/20
    Each
    Each - $25.84
    $25.84
    Each
       $25.84
     : 071.470.09
  • 50 ml
    24/40
    Each
    Each - $27.13
    $27.13
    Each
       $27.13
     : 071.470.10A
  • 125 ml
    19/22
    Each
    Each - $31.86
    $31.86
    Each
       $31.86
     : 071.470.51A
  • 125 ml
    24/40
    Each
    Each - $33.52
    $33.52
    Each
       $33.52
     : 071.470.14A