Round Bottom Flask, Straight, Triple Neck, Glassco
Supplier: Glassco
Certificates
About this item
Round bottom flask with joints, made from Borosilicate 3.3 glass
Heavy wall design. All products listed below are hand blown from tubing to ensure uniform wall thickness.
Frequently Bought Together