Quaternary Ammonium Compounds Method: Colorimetric With Test Strips 10 - 25 - 50 - 100 - 250 - 500 mg/L MQuant™, MilliporeSigma
Catalog # EM1.17920.0001
Supplier: MilliporeSigma
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Quaternary Ammonium Compounds Test
- Environmentally Preferable:
- Cat. No.:EM1.17920.0001
- Range:10-25-50-100-250-500 mg/L
- Supplier no.:1.17920.0001
- No. of tests:100 strips
Specifications
About this item
The test strip can be used to check quaternary ammonium compounds, which have microbicidal properties and are mainly used in surface cleaners.