ContactUsLinkComponent
Blank Test Strips Without Reagents MQuant™, MilliporeSigma
Blank Test Strips Without Reagents MQuant™, MilliporeSigma
  EM1.11860.0001
 :  
Blank Test Strips Without Reagents MQuant™, MilliporeSigma
  EM1.11860.0001
 :  1.11860.0001
 :  
  • Description:
    Blank Test Strips
  • Cat. No.:
    EM1.11860.0001
  • Supplier no.:
    1.11860.0001
  • No. of tests:
    100 strips
MQuant™; test strips are suitable for the semiquantitative detection of ions and of inorganic and organic substances and can easily be used anywhere.