Peracetic Acid Test Method: Colorimetric With Test Strips 5 - 10 - 20 - 30 - 50 mg/L MQuant™, MilliporeSigma
Catalog # EM1.10084.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Chemical method:Redox reaction
- Description:Test with Strips
- Cat. No.:EM1.10084.0001
- Range:5-10-20-30-50 mg/l CH₃CO₃H
- Supplier no.:1.10084.0001
- No. of tests:100 strips
Specifications
About this item
The test is suitable for the selective determination of peracetic acid traces in aqueous solutions and for check the absence of peracetic acid after rinsing processes, also in cases in which hydrogen peroxide is present.
MilliporeSigma # 1.10084.0001 replaces and is identical to MilliporeSigma # 10084-1