ContactUsLinkComponent
Nitrate Test RQeasy® Method: Reflectometric 5 - 250 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Nitrate Test RQeasy® Method: Reflectometric 5 - 250 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Catalog # EM1.17961.0001
Supplier:  MilliporeSigma
CAS Number:  
Nitrate Test RQeasy® Method: Reflectometric 5 - 250 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Catalog # EM1.17961.0001
Supplier:  MilliporeSigma
Supplier Number:  1.17961.0001
CAS Number:  
Specifications
  • Chemical method:
    Griess' reaction
  • Description:
    Test with Strips
  • Cat. No.:
    EM1.17961.0001
  • Range:
    5 - 250 mg/L NO₃
  • Supplier no.:
    1.17961.0001
  • No. of tests:
    50 strips
Specifications
About this item
The rapid determination of the nitrate content is of particular interest with fruit and vegetables to help decide whether a given delivery batch can be released for use in a specific product (e.g baby foods).
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.