Chlorine Test Method: Colorimetric With Test Strips 0 - 25 - 50 - 100 - 200 - 500 mg/L Cl₂ MQuant™, MilliporeSigma
Catalog # EM1.17924.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Chemical method:Redox reaction
- Description:Test with Strips
- Cat. No.:EM1.17924.0001
- Range:25-50-100-200-500 mg/l Cl₂
- Supplier no.:1.17924.0001
- No. of tests:100 strips
Specifications
About this item
The chlorine test is suitable for rapid determination of higher concentrations of chlorinated disinfectants, which are chiefly used in the food industry.