Vacuum #15 O-Ring Joint Tee Adapter with #15 Ace-Safe Connector Side Port and ½" Hose Barb, Ace Glass Incorporated
Catalog # 89428-018
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Vacuum #15 O-Ring Joint Tee Adapter with #15 Ace-Safe Connector Side Port and ½" Hose Barb
- Cat. No.:89428-018
- Supplier no.:8876-22
Specifications
About this item
This borosilicate glass adapter features two end 15mm O-Ring joints and a #15 Ace-Safe connector side port.
- Two end o-ring joints
- Side port
- Fabricated from borosilicate glass
Adapter is ideal for connecting components for Schlenk lines. Made mainly for connecting the Shlenk or vacuum line to a vacuum gauge. Ace Safe has ½" ID hose connection for connecting to vacuum tubing.