VWR® General Purpose Refrigerator Freezer Combination Units
Supplier: VWR International
Certificates
About this item
General-purpose, upright laboratory refrigerator/freezer combination units feature a mechanical dial thermostat, leveling legs, integrated handles, interior lights, and probe access port.
- Features heavy-duty reinforced handle and a magnetic door gasket for a tight seal
A spacious capacity allows for maximum storage space and flexibility for all types of materials. Unit operates with CFC–free refrigerant, foam insulation, and a mechanical thermostat.
Ordering information: Interior dimensions are subject to change without notice. Refrigerators and Freezers are supplied with a two-year parts and labor warranty, with an additional three-year warranty on compressor parts.