Order Entry
United States
ContactUsLinkComponent
HiTrap Columns, Capto Q, Cytiva
HiTrap Columns, Capto Q, Cytiva
Supplier:  Cytiva
Certificates
undefined
HiTrap Columns, Capto Q, Cytiva
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 95055-922
  • 95056-218
  • Max. Pressure:
  • 5 bar [0.5 MPa] (70 psi)
  • 5 bar [0.5 MPa] (70 psi)
  • pH Range:
  • 2 - 12
  • 2 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Frequently Bought Together