Order Entry
United States
ContactUsLinkComponent
HiScreen Octyl FF Columns
HiScreen Octyl FF Columns
Catalog # 97067-810
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
HiScreen Octyl FF Columns
Catalog # 97067-810
Supplier:  Cytiva
Supplier Number:  28-9269-86
Product available on GSA Advantage®

Specifications

  • Max. Pressure:
    1.5 bar [0.15 MPa] (22 psi)
  • Capacity:
    4.7 ml
  • Storage:
    4 to 30 °C, 20% Ethanol
  • Column hardware:
    PP
  • Bed dimensions:
    7.7×100 mm
  • Flow velocity:
    < 240 cm/h
  • Cat. No.:
    97067-810
  • Supplier no.:
    28926986

Specifications

About this item

HiScreen Octyl FF columns are prepacked with Octyl Sepharose 4 Fast Flow hydrophobic interaction chromatography (HIC) resins. The columns are an excellent choice for method optimization and parameter screening.

  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same resin using the same linear fluid velocity

Octyl Sepharose 4 Fast Flow is a well-established, standard aliphatic HIC resin for capture and intermediate purification.

The small column volume of 4.7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.

If necessary, two columns can easily be connected in series to give a bed height of 20 cm.