Order Entry
United States
ContactUsLinkComponent
 
HiPrep Desalting Columns, Cytiva
 :  Cytiva
 
undefined
HiPrep Desalting Columns, Cytiva
 :  Cytiva
 

 

  • Catalog No:
  • 95055-814
  • 95055-870
  • Description:
  • HiPrep™ 26/10 Desalting
  • HiPrep™ 26/10 Desalting
  • Max. Pressure:
  • 1.5 bar [0.15 MPa] (22 psi)
  • 1.5 bar [0.15 MPa] (22 psi)
  • pH Range:
  • 2 - 13
  • 2 - 13
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

 

Frequently Bought Together