StrepTactin™ Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Avantor
Certificates
About this item
StrepTactin Sepharose High Performance media for robust, high resolution purification of Strep II-tagged proteins
- Highly pure Strep II-tagged recombinant proteins eluted in concentrated form and small volumes
- Physiological conditions and mild elution preserve target protein activity
- Fast and easy regeneration with 0.5 M NaOH
- Compatible with a wide range of reducing agents, detergents, denaturants and other additives
- Stable in all commonly used buffers, 0.5 M NaOH (regeneration and cleaning), reducing agents, denaturants, and detergents
Mild purification conditions preserve antigenicity and function. This robust and stable Sepharose based medium results in high resolution, sharp concentrated peaks, and reproducible purifications. The binding capacity is also good. Binding capacity is protein dependent. Dynamic binding capacity was tested with GAPDH-Strep(II), Mr 37 400.
Specifications
- Matrix:Highly cross-linked agarose, 6%
- Ligand:StrepTactin
- Storage Temperature:2 - 8 °C
- Binding Capacity:6 mg Strep II-tagged protein/ml
- Flow Rate:<300 cm/h
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together