MAbTrap Column Kit, Cytiva
Catalog # 95056-072
Supplier: Cytiva
Product available on GSA Advantage®
Specifications
- Flow rate:< 4 mL/min
- Particle Size:34 µm
- Max. Pressure:3 bar [0.3 MPa] (42 psi)
- Capacity:1 ml
- Description:MAbTrap™ kit
- pH Range:3 - 9
- Column hardware:PP
- Bed dimensions:7×25 mm
- Cat. No.:95056-072
- Supplier no.:17-1128-01
Specifications
About this item
MAbTrap Kit is an affinity chromatography kit for fast and effective purification of monoclonal and polyclonal IgG from ascites fluid, serum, and cell culture supernatants.
- Convenient kit for purification of IgG from monoclonal and polyclonal material using a syringe in less than 15 min
- MAbTrap Kit contains everything necessary to perform a complete one-step purification without using complex equipment.
MAbTrap Kit contains everything necessary to perform a complete one step purification without using complicated equipment. It also eliminates time-consuming buffer preparation and column packing. The kit includes one HiTrap column, prepacked with Protein G Sepharose High Performance and enough buffer concentrate for 20 preparations with a syringe.