Order Entry
United States
ContactUsLinkComponent
His GraviTrap TALON Columns, Cytiva
His GraviTrap TALON Columns, Cytiva
Catalog # 11010-720
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
His GraviTrap TALON Columns, Cytiva
Catalog # 11010-720
Supplier:  Cytiva
Supplier Number:  29-0005-94
Product available on GSA Advantage®

Specifications

  • Capacity:
    1 ml
  • Description:
    His GraviTrap™ TALON®
  • Binding Capacity:
    up to 15mg histidine-tagged protein
  • Column hardware:
    PP
  • Cat. No.:
    11010-720
  • Supplier no.:
    29000594

Specifications

About this item

His GraviTrap TALON® gravity flow columns are designed for convenient and efficient gravity flow purification of histidine-tagged proteins directly from clarified or unclarified cell lysates.

  • Convenient histidine-tagged purification with high purity (> 90%) and binding capacity of up to 15 mg/column
  • Ready-to-use gravity flow column prepacked with 1 ml TALON® Superflow™ cobalt-IMAC medium
  • Offering a different selectivity compared to nickel-charged media
  • One run takes about 30 minutes, and large sample volumes can be applied all at once

Large sample volumes can be applied in one go, and the histidine-tagged protein is effectively eluted in a small volume. LabMate PD-10 Buffer Reservoir can be connected to the columns for handling of sample volumes above 10 ml. Leakage of cobalt ions from TALON® Superflow™ medium is negligible.

His GraviTrap TALON® columns are delivered in a package that can be converted into a column stand to simplify purification, eliminating the need for additional equipment to complete the protocol. Special column frits protect the chromatography medium from running dry during purification.