ContactUsLinkComponent
Collection Plates for 96-Well Plates, Cytiva
Collection Plates for 96-Well Plates, Cytiva
Catalog # 95056-282
Supplier:  Cytiva
Product available on GSA Advantage®
undefined
Collection Plates for 96-Well Plates, Cytiva
Catalog # 95056-282
Supplier:  Cytiva
Supplier Number:  28-4039-43
Product available on GSA Advantage®
Specifications
  • Well volume:
    <500 µL
  • Description:
    Collection Plates
  • Cat. No.:
    95056-282
  • Supplier no.:
    28-4039-43
Specifications
About this item
Made from biocompatible PP. Collection plates, 96-well, designed for collection of eluted samples in parallel from screening plates such as MultiTrap™ and PreDictor™ plates.
  • Convenient collection of samples in 96-well format
  • Excellent fit to prepacked MultiTrap plates and PreDictor plates
  • V-bottom for minimal loss of sample
  • Collection of < 500µL volumes of sample per well