ContactUsLinkComponent
PIG® HazMat Spill Kit in Large Wall-Mount Cabinet, New Pig
PIG® HazMat Spill Kit in Large Wall-Mount Cabinet, New Pig
Catalog # 56620-488
Supplier:  New Pig
undefined
PIG® HazMat Spill Kit in Large Wall-Mount Cabinet, New Pig
Catalog # 56620-488
Supplier:  New Pig
Supplier Number:  KIT328
Specifications
  • Capacity:
    Up to 68.1 L (18 gal.)
  • Cat. No.:
    56620-488
  • Supplier no.:
    KIT328
Specifications
About this item
Safety-yellow, heavy-gauge, welded steel cabinet is printed "Spill Kit" for quick identification
Prepacked cabinet provides easy access to contents. See-through front panel lets you easily check inventory levels. Includes write-on label for emergency contact info. Two adjustable shelves.
PIG HazMat Socks stop spreading spills; PIG HazMat Pads and Pillows absorb; PIG Repair Putty plugs and patches cracks and leaks. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide