ContactUsLinkComponent
PIG® Spill Kit in 95-Gallon Wheeled Overpack Salvage Drum, New Pig
PIG® Spill Kit in 95-Gallon Wheeled Overpack Salvage Drum, New Pig
  56620-410
 :  New Pig
undefined
PIG® Spill Kit in 95-Gallon Wheeled Overpack Salvage Drum, New Pig
  56620-410
 :  New Pig
 :  KIT241-WH
  • Capacity:
    Up to 227.1 L (60 gal.)
  • Cat. No.:
    56620-410
  • Supplier no.:
    KIT241-WH
Overpack drum container is UN Rated for shipping waste after spill cleanup
  • Temporary disposal bags help make cleanup easier
  • Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail
  • Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry
  • Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole
  • Large, color-coded label makes identification fast and accurate
  • Ledges molded into container make kit easy to move by forklift
  • Tamperproof seals help prevent pilfering of spill response supplies
6" polyolefin wheels. UN1H2/X113/S.
Lift-out, prepacked baskets speed access and guard contents from UV. PIG Blue Socks stop spreading spills; PIG Mat and Pillows absorb; wipers clean surfaces and hands