ContactUsLinkComponent
PIG® Spill Kit in Bucket, New Pig
PIG® Spill Kit in Bucket, New Pig
Catalog # 56620-414
Supplier:  New Pig
undefined
PIG® Spill Kit in Bucket, New Pig
Catalog # 56620-414
Supplier:  New Pig
Supplier Number:  KIT213
Specifications
  • Capacity:
    Up to 15.1 L (4 gal.)
  • Environmentally Preferable:
  • Cat. No.:
    56620-414
  • Supplier no.:
    KIT213
Specifications
About this item
Polyethylene bucket with easy-open, screw-on lid can be used to ship or temporarily store used absorbents
  • Prepacked container provides easy access to contents
  • Large, color-coded label makes identification fast and accurate
  • Tamper-proof seals help prevent pilfering of spill response supplies
  • Use as refill for wall-mount holder (sold separately)
  • No wheels
PIG BLUE Socks stop spreading spills; PIG Mat absorbs quickly.
Certifications: UN Rating 1H2/Y30/S for Packing Groups II and III.