ContactUsLinkComponent
PIG® HazMat Chemical Absorbent Pulp, New Pig
PIG® HazMat Chemical Absorbent Pulp, New Pig
Catalog # 56620-186
Supplier:  New Pig
undefined
PIG® HazMat Chemical Absorbent Pulp, New Pig
Catalog # 56620-186
Supplier:  New Pig
Supplier Number:  HA8010
Specifications
  • Description:
    HazMat Chemical Absorbent Pulp, Absorbs Acids, Bases and Unknowns, One 5 lb. Bag
  • Cat. No.:
    56620-186
  • Supplier no.:
    HA8010
Specifications
About this item
Packing the power of PIG HazMat in a loose absorbent, this pulp can be used in a variety of applications
  • Absorbs and retains most acids, bases and unknown liquids
  • Pink color is easily distinguishable to assure workers use the correct pulp during a spill emergency
  • Can be incinerated after use to reduce waste
Absorbs the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant pulp won't degrade or cause a dangerous reaction upon contact with corrosive spills. Excellent for difficult to reach spaces or final cleanup