ContactUsLinkComponent
PIG® Plug-N-Seal® Patching Paste, New Pig
PIG® Plug-N-Seal® Patching Paste, New Pig
Catalog # 89205-886
Supplier:  New Pig
undefined
PIG® Plug-N-Seal® Patching Paste, New Pig
Catalog # 89205-886
Supplier:  New Pig
Supplier Number:  PTY220
Specifications
  • Description:
    Patching Paste for Fuel Tanks, Steel Drums, and Containers, 1 lb. Container
  • Environmentally Preferable:
  • Cat. No.:
    89205-886
  • Supplier no.:
    PTY220
Specifications
About this item
Temporarily stops leaks until system can be drained or leak source contained
No preparation needed, even on dirty or rusty surfaces. Temporarily stops leaks from 2 minutes up to 2 hours. Ready to use without mixing.
Excellent chemical resistance makes it excellent for flammable or hazardous leaks.