ContactUsLinkComponent
PIG® 4-in-1® Oil-Only Absorbent Mat Roll, New Pig
PIG® 4-in-1® Oil-Only Absorbent Mat Roll, New Pig
Catalog # 89205-924
Supplier:  New Pig
undefined
PIG® 4-in-1® Oil-Only Absorbent Mat Roll, New Pig
Catalog # 89205-924
Supplier:  New Pig
Supplier Number:  MAT435
Specifications
  • Description:
    Mediumweight
  • Dimensions:
    40.6 cm x 46 m (16" x 150')
  • Cat. No.:
    89205-924
  • Supplier no.:
    MAT435
Specifications
About this item
Exclusive 4-in-1 Mat is a pad, roll, sock and wiper all in one
Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water. Flame-resistant material melts when exposed to high heat. Rolls are perforated every 10"; easy to tear to size, use only what you need.
Mediumweight construction is versatile for general, everyday tasks and cleanups.
Use as a pad to catch drips and soak up oily spills; roll out to cover large areas; fold along score lines to make a thick, absorbent "sock"; tear off a piece to use as a low-lint wiper. No matter how you use it, this mat builds on the proven eight-layer, 100% polypropylene construction of PIG Mat. Scuff-resistant, spunbond outer layer resists wear and tear, even under heavy foot traffic or abrasive conditions. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue