Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant IL-15 Mutant H:Fc Ms B 1Vial (from CHO cells), Biotin
Catalog # 102513-880
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Human Recombinant IL-15 Mutant H:Fc Ms B 1Vial (from CHO cells), Biotin
Catalog # 102513-880
Supplier:  Adipogen
Supplier Number:  CHI-HF-21115MBI-1
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Biotin
  • Protein Function:
    Interleukins
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    1 Vial
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Biological Activity:
    This mutant IL-15/Fc fusion protein specifically binds to the IL-15R, competitively inhibits IL-15-triggered cell proliferation, promotes transplant tolerance, does not activate the STAT-signaling pathway, and exerts a prolonged circulating half-life caused by the modified Fc domain.
  • Gene ID:
    NP_751914.1
  • Protein Synonyms:
    Interleukin-15
  • Protein/Peptide Name:
    IL-15 Mutant H:Fc Ms B 1Vial
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized. In PBS and 0.09% sodium azide.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Tested Applications:
    Immunofluorescent staining, Flow cytometry
  • Cat. No.:
    102513-880
  • Supplier no.:
    CHIHF21115MBI1

Specifications

About this item

Interleukin-15 (IL-15) has a broad spectrum of biological activities. It is crucial for the development, proliferation, survival and differentiation of multiple cells from both innate and adaptive immune systems. IL-15 up-regulation has a central role in the development of several autoimmune or chronic inflammatory disorders. Targeting IL-15 or its receptor may have a valuable impact on the treatment of immune-mediated diseases. IL-15 participates in the development of important immune antitumor mechanisms. It activates CD8(+) T cells, natural killer (NK) cells, NK T cells, and can promote the formation of antitumor antibodies. IL-15 can also protect T effector cells from the action of T regulatory cells and reverse tolerance to tumor-associated antigens. In pre-clinical studies IL-15 has been found to demonstrate potentiated antitumor effects following pre-association with IL-15Ralpha, or when used in combination with chemotherapy, adoptive therapy, monoclonal antibodies, and tumor vaccines. Application: Useful for immunofluorescent staining and flow cytometric analysis to identify and enumerate IL-15Ralpha expressing cells within mixed cell populations.