To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:50 µg
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Biological Activity:This mutant IL-15/Fc fusion protein specifically binds to the IL-15R, competitively inhibits IL-15-triggered cell proliferation, promotes transplant tolerance, does not activate the STAT-signaling pathway, and exerts a prolonged circulating half-life caused by the modified Fc domain.
- Gene ID:NP_751914.1
- Protein Synonyms:Interleukin-15
- Protein/Peptide Name:IL15
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice
- Cat. No.:102511-356
- Supplier no.:HF21015MC050
Interleukin-15 (IL-15) has a broad spectrum of biological activities. It is crucial for the development, proliferation, survival and differentiation of multiple cells from both innate and adaptive immune systems. IL-15 up-regulation has a central role in the development of several autoimmune or chronic inflammatory disorders. Targeting IL-15 or its receptor may have a valuable impact on the treatment of immune-mediated diseases. IL-15 participates in the development of important immune antitumor mechanisms. It activates CD8(+) T cells, natural killer (NK) cells, NK T cells, and can promote the formation of antitumor antibodies. IL-15 can also protect T effector cells from the action of T regulatory cells and reverse tolerance to tumor-associated antigens. In pre-clinical studies IL-15 has been found to demonstrate potentiated antitumor effects following pre-association with IL-15Ralpha, or when used in combination with chemotherapy, adoptive therapy, monoclonal antibodies, and tumor vaccines.