Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant IL6 (from CHO Cells)
Mouse Recombinant IL6 (from CHO Cells)
Supplier:  Adipogen
Certificates
Mouse Recombinant IL6 (from CHO Cells)
Supplier:  Adipogen

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Interleukins
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Biological Activity:
    Shows the biological function of the IL-6 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
  • Gene ID:
    NP_112445.1
  • Protein Synonyms:
    Interleukin-6|B Cell Hybridoma Growth Factor|Interleukin HP-1
  • Protein/Peptide Name:
    IL6
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice

Specifications

Frequently Bought Together