About this item
Interleukin-4 (IL-4) is a cytokine produced by type 2 helper T cells, the Th2 cells. These cells tends to make a specific set of lymphokines including IL-4, IL-5, IL-6, IL-10, IL-13, IL-3 and GM-CSF and fail to produce IL-2, IFN-gamma, and lymphotoxin (TNF-beta). In addition, mast cells can produce IL-4. IL-4 exerts numerous effects on various hematopoietic cell types. On B cells, IL-4 promotes immunoglobulin class switching to IgE and IgG1 isotypes and upregulates MHC class II and CD23 expression. IL-4 promotes survival, growth, and differentiation of both T and B lymphocytes, mast cells and endothelial cells. In addition, IL-4 inhibits the production of TNF, IL-1, and IL-6 by macrophages.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein Function:Interleukins
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:<0.06EU/µg
- Biological Activity:Shows the biological function of the IL-4 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
- Gene ID:NP_067258.1
- Protein Synonyms:B Cell Stimulatory Factor 1|BSF-1|Interleukin-4
- Protein/Peptide Name:IL4
- Purity:98%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.2µm-filtered solution in PBS.
- Shipping Temperature:-20 °C, Blue Ice