Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD40 Ligand (from CHO Cells)
Human Recombinant CD40 Ligand (from CHO Cells)
Catalog # 102511-342
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Human Recombinant CD40 Ligand (from CHO Cells)
Catalog # 102511-342
Supplier:  Adipogen
Supplier Number:  CHI-HF-210CD40L-C050
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_000065.1
  • Protein Synonyms:
    CD154|TNFSF5|Tumor Necrosis Factor Ligand Superfamily Member 5|Gp39
  • Protein/Peptide Name:
    CD40 Ligand
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102511-342
  • Supplier no.:
    HF210CD40LC050

Specifications

About this item

CD40 Ligand (CD40L), renamed TNFSF5 but now also known as CD154, TRAP and gp39, is a 34-39kDa type II transmembrane glycoprotein that belongs to the TNF superfamily. As with other TNF superfamily members, CD40L will exist as a trimer, both as a membrane bound and soluble form. Multiple mutations and alternate splice forms of CD40L exist, often associated with pathology and leading to truncated or nontrimerizable forms of CD40L. CD40L binds to both CD40 and to integrin alphaIIbbeta3 (CD41). In the cell membrane, it also associates with a unique splice variant of CD28 (CD28i) that may facilitate CD40L signal transduction. CD40L is expressed by monocytes, NK cells, mast cells, basophils, smooth muscle cells, endothelial cells, dendritic cells,activated and resting B cells, plus activated platelets and CD4+ T cells. CD40L ligation of CD40 on dendritic cells (DC) initiates DC maturation and differentiation. CD40L signaling into naive B cells promotes germinal center formation and isotope switching. CD40-CD40L seems to bridge innate and adaptive immune signals.