Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant CD275 (B7-H2) (Ms):Fc (from CHO cells)
Mouse Recombinant CD275 (B7-H2) (Ms):Fc (from CHO cells)
  102511-336
 :  Adipogen
Small Business Enterprise
 :  
Mouse Recombinant CD275 (B7-H2) (Ms):Fc (from CHO cells)
  102511-336
 :  Adipogen
 :  CHI-MF-110B7H2-C100
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.06EU/µg
  • Gene ID:
    NP_056605.1
  • Protein Synonyms:
    ICOS Ligand|B7-H2|B7RP-1|B7-like Protein Gl50
  • Protein/Peptide Name:
    CD275 (B7-H2) (Ms):Fc
  • Purity:
    98%
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.2µm-filtered solution in PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102511-336
  • Supplier no.:
    MF110B7H2C100

 

 

Mouse B7-H2, also called CD275, B7RP-1, B7h, LICOS and GL50, is a member of the growing B7 family of immune costimulatory proteins. CD275 has been identified as the ligand for ICOS, a member of the CD28 family of co-stimulatory receptors. Mouse CD275 is expressed on resting B cells and at low levels on monocytes. The CD275/ ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.