Order Entry
United States
Orders LinkContactUsLinkComponent
Strobilurin B ≥97%

Strobilurin B ≥97%

Irritant
Marketsource
Supplier:   Adipogen
Strobilurin B ≥97%
Supplier:  Adipogen
Marketsource
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Antifungal. Respiration inhibitor. Blocks the electron transfer between cytochrome b and cytochrome c1.

  • CAS Number:65105-52-4
  • Formula:C₁₇H₁₉ClO₄
  • Molecular Weight:322.8 g/mol
  • Storage Temperature:Freezer
Specifications for Product Family

Specification Test Results

  • Source/Host:
    Isolated from Strobilurus sp.
  • Purity:
    97% (HPLC)
  • Appearance/Color:
    Light brown solid.
  • Solubility:
    Soluble in DMSO or methanol.
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic vial
    500 µg
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $173.08
    $173.08
    Each (500µG)
       $173.08

    Supplier Number: BVT-0312-C500
    Specifications:
    Marketsource
    Marketsource
  • Plastic vial
    1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $276.92
    $276.92
    Each (1mg)
       $276.92

    Supplier Number: BVT-0312-M001
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.