Order Entry
United States
ContactUsLinkComponent
 
(-)-Ageloxime D ≥97% (by HPLC)
Catalog # 102515-716
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
(-)-Ageloxime D ≥97% (by HPLC)
Catalog # 102515-716
Supplier:  Adipogen
Supplier Number:  AG-CN2-0016-M001
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Size:
    1 mg
  • Cat. No.:
    102515-716
  • Supplier no.:
    AGCN20016M001

Specifications

Specification Test Results

  • Source/Host:
    Isolated from sponge Agelas sp.
  • Purity:
    97% (HPLC)
  • Appearance/Color:
    Beige solid.
  • Solubility:
    Soluble in ethanol or DMSO.

Specification Test Results

About this item

 :
 :438.6 g/mol
Storage Temperature:Freezer
CAS Number:1219817-25-0
Formula:C₂₆H₄₀N₅O