Order Entry
United States
ContactUsLinkComponent
RELM-beta (human) Matched Pair Detection Set
RELM-beta (human) Matched Pair Detection Set
Catalog # 102515-632
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
RELM-beta (human) Matched Pair Detection Set
Catalog # 102515-632
Supplier:  Adipogen
Supplier Number:  AG-46A-0001-KI01
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    RELM-β
  • Description:
    RELM-beta (human) Matched Pair Detection Set
  • Size:
    1 Set
  • Storage Conditions:
    -20°C (long term); +4°C (short term)
  • Sample Type:
    Serum
  • Cross Reactivity:
    Does not cross-react with human adiponectin, mouse adiponectin, human RBP4, human Nampt, human vaspin, human GPX3, human ANGPTL3, human progranulin, human leptin, human resistin, human IL-33, human sirtuin 1, human clusterin, mouse resistin, human FABP4.
  • Detection Method:
    Colorimetric
  • Shelf Life:
    Stable for at least 6 months after receipt when stored at +4°C.
  • Detection Range:
    0.016 to 1ng/ml
  • Sensitivity:
    10 pg/ml
  • Cat. No.:
    102515-632
  • Supplier no.:
    AG46A0001KI01
  • No. of tests:
    Reagents for 5× 96-well plates

Specifications

About this item

RELM-beta is a 19kDa disulfide-linked homodimeric protein expressed in the epithelium of the colon and small bowel

RELM-beta has been suggested to play a regulatory role during inflammation and may also act to establish links among adipose tissue, the intestine and the liver. Interestingly, the molecular structure of RELM-beta is highly homologous to that of the adipose-derived cytokine resistin. The specific and sensitive RELM-beta Matched Pair Detection Set might detect RELM-beta in serum of gut-inflamed patients (e.g. inflammatory bowel diseases, Crohn disease and ulcerative colitis). Natural levels of RELM-beta were seen in stool, but were not detectable in the serum or other cell culture models of healthy human patients. Further testing are ongoing.