Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant Megaox40L (from HEK293 cells)
Mouse Recombinant Megaox40L (from HEK293 cells)
Catalog # 102515-470
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Mouse Recombinant Megaox40L (from HEK293 cells)
Catalog # 102515-470
Supplier:  Adipogen
Supplier Number:  AG-40B-0029-C010
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    10 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <0.01EU/µg
  • Biological Activity:
    Activates T cell proliferation.
  • Gene ID:
    P43488
  • Protein Synonyms:
    ACRP30headless:OX40L|ACRP30headless:CD134L|ACRP30headless:CD252|MultimericOX40L™
  • Protein/Peptide Name:
    Megaox40L
  • Purity:
    90%
  • Molecular Weight:
    ~40kDa (SDS-PAGE)
  • Sequence:
    18-111
  • Endotoxin Level:
    Low
  • Concentration:
    0.1mg/ml after reconstitution.
  • Formulation:
    Lyophilized. Contains PBS.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102515-470
  • Supplier no.:
    AG40B0029C010

Specifications

About this item

OX40L is a membrane-expressed cytokine that belongs to the TNF ligand family. It acts as costimulator through interaction with its ligand OX40 on T cells, stimulating T cell activation, proliferation and cytokine production. OX40L is expressed on antigen presenting cells including B cells, dendritic cells, mast cells and endothelium. MultimericOX40L is a high activity construct in which two trimeric OX40L ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively activates T cell proliferation.