About this item
MultimericCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo. It provides a simple and equally potent alternative to combinations. MultimericCD40L™ has shown to suppress alum-induced IL-1beta release and caspase-1 activation in a dose-, CD40- and time dependent manner, without affecting BMDM viability. It also effectively suppressed the inflammasome function triggered by NLRP3 activators. The secretion of caspase-1 independent inflammatory mediators has been shown to be unaltered or even enhanced.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<0.01EU/µg
- Biological Activity:Induces B cells activation (as demonstrated by dose-dependent upregulation of CD86) (ED50: <1ng/ml).
- Gene ID:P29965
- Protein Synonyms:ADIPOQ-CD40L|ACRP30headless:CD154|ACRP30headless:TNFSF5|MultimericCD40L™|ACRP30headless:CD40L
- Protein/Peptide Name:CD40L
- Purity:95%
- Molecular Weight:~35-40kDa (SDS-PAGE)
- Sequence:18-111
- Endotoxin Level:Low
- Concentration:0.1mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS
- Shipping Temperature:-20 °C, Blue Ice