About this item
Tumor necrosis factor (TNF, cachexin or cachectin and formerly known as tumor necrosis factor-alpha) is a cytokine involved in systemic inflammation and is a member of a group of cytokines that stimulate the acute phase reaction. The receptor TNF-R1 is activated by both the membrane-bound and soluble trimeric forms of TNF-alpha, whereas the receptor TNF-R2 only responds to the membrane-bound form of TNF-alpha (MultimericTNF-alpha (Prod. No. AG-40B-0019). Since the MultimericTNF-alpha mimics the membrane-bound form (forms oligomers higher than trimer), it is the only TNF-alpha protein that can activate the TNF-R2. For TNF-R1 activation, either "normal" TNF-alpha or MultimericTNF-alpha can be used.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:<0.01EU/µg
- Biological Activity:Induces cell death at a concentration range of 0.01-10ng/ml (ED50= 0.05ng/ml or 2 x 10E7 Units/mg; WEHI 164 cells).
- Gene ID:P01375
- Protein Synonyms:Cachectin|Tumor Necrosis Factor Ligand Superfamily Member 2
- Protein/Peptide Name:TNF alpha (soluble)
- Purity:95%
- Molecular Weight:~17kDa (SDS-PAGE)
- Sequence:85-223
- Endotoxin Level:Low
- Concentration:10 µg size: 0.1 mg/ml after reconstitution.50 µg size: 1 mg/ml after reconstitution.
- Formulation:Lyophilized. Contains PBS.
- Shipping Temperature:-20 °C, Blue Ice