Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant ST2 (from HEK293 Cells)
  102514-884
 :  Adipogen
Small Business Enterprise
 :  
Human Recombinant ST2 (from HEK293 Cells)
  102514-884
 :  Adipogen
 :  AG-40A-0059-C050
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    50 µg
  • Environmentally Preferable:
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <5EU/mg
  • Biological Activity:
    Interacts with human IL-33.
  • Gene ID:
    Q01638
  • Protein Synonyms:
    IL-1 Receptor-like 1|IL-33 Receptor|Interleukin-33 Receptor|Interleukin-1 Receptor-like 1|IL1RL1
  • Protein/Peptide Name:
    ST2
  • Purity:
    90%
  • Molecular Weight:
    ~90kDa (SDS-PAGE)
  • Sequence:
    1-328
  • Endotoxin Level:
    Low
  • Concentration:
    ~0.5mg/ml
  • Formulation:
    Liquid. 0.2µm-filtered solution in PBS, pH 7.2.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102514-884
  • Supplier no.:
    AG40A0059C050

 

 

The ST2 (Interleukin-1 receptor-like 1; Interleukin-33 receptor) gene was originally identified as a gene induced by serum or oncogene expression in fibroblasts. The gene produces a shorter soluble secreted form (ST2) and a longer, transmembrane form (ST2L) by alternative splicing. Soluble ST2 has been shown to downregulate the expression of TLR1 and TLR4. ST2L negatively regulates TLR4 signaling and induces endotoxin tolerance, and enhances Th2 responses. IL-33 is the specific ligand for ST2L.