Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant Clusterin (from E. coli)
Mouse Recombinant Clusterin (from E. coli)
Catalog # 102514-878
Supplier:  Adipogen
Small Business Enterprise
CAS Number:  
Mouse Recombinant Clusterin (from E. coli)
Catalog # 102514-878
Supplier:  Adipogen
Supplier Number:  AG-40A-0057-C050
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <5EU/mg
  • Gene ID:
    Q549A5
  • Protein Synonyms:
    Apolipoprotein J|SGP-2|TRPM-2|CLI|APO-J|CLU
  • Protein/Peptide Name:
    Clusterin
  • Purity:
    90%
  • Molecular Weight:
    ~45kDa (SDS-PAGE)
  • Sequence:
    22-448
  • Endotoxin Level:
    Low
  • Concentration:
    1mg/ml
  • Formulation:
    Liquid. 0.2µm-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl.
  • Shipping Temperature:
    -20 °C, Blue Ice
  • Cat. No.:
    102514-878
  • Supplier no.:
    AG40A0057C050

Specifications

About this item

Clusterin shares homology with the small heat shock protein family of molecular chaperones. The mature secreted form of the protein is a glycosylated, 80-kDa disulfide-linked heterodimer of alpha and beta subunits (produced by internal cleavage). Clusterin is expressed in virtually all tissues and found in all human fluids. It is involved in numerous physiological processes important for carcinogenesis and tumor growth, including apoptotic cell death, cell cycle regulation, DNA repair, cell adhesion, tissue remodeling, lipid transportation, membrane recycling, and immune system regulation. Clusterin also exists as a nuclear protein. The secreted form of Clusterin has extracellular chaperone and anti-apoptotic activities while the nuclear form acts as a proapoptotic factor.